Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tert | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009354.1 | NP_033380.1 | 9 - 10 | 2585 | 84 | |
GenBank mRNA | AF051911.1 | - | 9 - 10 | 2585 | 84 | |
AF073311.1 | - | 9 - 10 | 2556 | 84 | ||
BC082327.1 | - | 10 - 11 | 2087 | 84 | ||
BC127068.1 | - | 10 - 11 | 1873 | 40110173 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |