Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Upf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122829.1 | NP_001116301.1 | 13 - 14 | 2117 | 63 | |
NM_030680.2 | NP_109605.2 | 13 - 14 | 2084 | 63 | ||
GenBank mRNA | AF182947.1 | - | 14 - 15 | 2073 | 63 | |
AF322655.1 | - | 13 - 14 | 1804 | 63 | ||
AK052517.1 | - | 2 - 3 | 189 | 63 | ||
AK148196.1 | - | 14 - 15 | 2094 | 63 | ||
BC030916.1 | - | 1 - 2 | 95 | 63 | ||
BC052149.1 | - | 13 - 14 | 2022 | 6332288 | 63 | |
BC056442.1 | - | 13 - 14 | 2117 | 5702493 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |