Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc9a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081060.1 | NP_001074529.1 | 5 - 6 | 919 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Transporter Mus musculus Probe spans exons |