Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arrdc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042591.1 | NP_001036056.1 | 3 - 4 | 743 | 124 | |
GenBank mRNA | AK084951.1 | - | 3 - 4 | 754 | 124 | |
AK088288.1 | - | 3 - 4 | 758 | 124 | ||
AK122499.1 | - | 3 - 4 | 730 | 124 | ||
AK163364.1 | - | 3 - 4 | 288 | 124 | ||
BC031125.1 | - | 1 - 2 | 121 | 124 | ||
BC044762.1 | - | 3 - 4 | 741 | 124 | ||
BC054844.1 | - | 3 - 4 | 743 | 6493441 | 124 | |
BC098474.1 | - | 3 - 4 | 737 | 30094818 | 124 | |
DQ861994.1 | - | 3 - 4 | 488 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |