Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cylc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162865.1 | NP_001156337.1 | 1 - 2 | 51 | 121 | |
GenBank mRNA | AK015651.1 | - | 1 - 2 | 52 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |