Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clcn4-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK035804.1 | - | 6 - 7 | 824 | 93 | |
AK088317.1 | - | 4 - 5 | 585 | 93 | ||
AK133759.1 | - | 6 - 7 | 850 | 93 | ||
AK160325.1 | - | 5 - 6 | 1424 | 93 | ||
BC005553.1 | - | 6 - 7 | 885 | 3586154 | 93 | |
BC110668.1 | - | 7 - 8 | 1184 | 6391053 | 93 | |
DQ186662.1 | - | 5 - 6 | 682 | 93 | ||
Z49916.1 | - | 7 - 8 | 1095 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |