Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rassf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028478.2 | NP_082754.2 | 6 | 572 | 67 | |
GenBank mRNA | BC039934.1 | - | 6 | 583 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |