Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agrn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021604.3 | NP_067617.3 | 19 - 20 | 3432 | 98 | |
GenBank mRNA | BC058651.1 | - | 18 - 19 | 3545 | 98 | |
BC059259.1 | - | 6 - 7 | 846 | 98 | ||
BC150703.1 | - | 17 - 18 | 2886 | 9087614 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |