Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssxb8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081565.3 | NP_001075034.3 | 3 - 4 | 281 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Mus musculus Probe spans exons |