Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK165793.1 | - | 5 | 542 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |