Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cmklr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008153.3 | NP_032179.1 | 1 - 2 | 170 | 85 | |
GenBank mRNA | BC100612.1 | - | 1 - 2 | 170 | 2654794 | 85 |
BY347978.1 | - | 1 - 2 | 207 | 85 | ||
U79525.1 | - | 1 - 2 | 172 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Mus musculus 1700 Probe spans exons |