Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnj4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008427.4 | NP_032453.3 | 1 - 2 | 56 | 84 | |
GenBank mRNA | BC120593.1 | - | 1 - 2 | 33 | 40129516 | 84 |
BC137594.1 | - | 1 - 2 | 33 | 8860613 | 84 | |
S71382.1 | - | 1 - 2 | 57 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |