Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim67 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK051455.1 | - | 1 | 3256 | 84 | |
AK220282.1 | - | 1 | 2701 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon |