Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snw1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025507.2 | NP_079783.2 | 7 - 8 | 752 | 69 | |
GenBank mRNA | AK009218.1 | - | 2 - 3 | 107 | 69 | |
AK012384.1 | - | 2 - 3 | 107 | 69 | ||
AK166163.1 | - | 7 - 8 | 753 | 69 | ||
AK167695.1 | - | 7 - 8 | 725 | 69 | ||
AX775977.1 | - | 7 - 8 | 727 | 69 | ||
AX775979.1 | - | 7 - 8 | 727 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |