Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lingo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001165999.1 | NP_001159471.1 | 1 - 2 | 665 | 133 | |
GenBank mRNA | AK043936.1 | - | 1 - 2 | 667 | 133 | |
AK139458.1 | - | 1 - 2 | 666 | 133 | ||
AK141568.1 | - | 1 - 2 | 148 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Mus musculus 1700 Probe spans exons |