Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Islr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195431.1 | NP_001182360.1 | 1 - 2 | 132 | 64 | |
GenBank mRNA | AB024538.1 | - | 1 - 2 | 85 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus 1700 Inventoried Probe spans exons |