Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pfdn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027044.3 | NP_081320.2 | 3 - 4 | 283 | 180 | |
GenBank mRNA | AF108357.1 | - | 3 - 4 | 243 | 180 | |
AK004438.1 | - | 3 - 4 | 238 | 180 | ||
AK005682.1 | - | 3 - 4 | 229 | 180 | ||
AK028206.1 | - | 3 - 4 | 220 | 180 | ||
AK145883.1 | - | 3 - 4 | 236 | 180 | ||
BC026920.1 | - | 3 - 4 | 220 | 3666296 | 180 | |
CN835522.1 | - | 3 - 4 | 286 | 180 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |