Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200010833.mm.1 |
NM_001145902.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200010833.mm.1 |
NM_001145902.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200010833.mm.1 |
NM_146124.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200010833.mm.1 |
NM_146124.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200010833.mm.1 |
NR_027371.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200010833.mm.1 |
NR_027371.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200010833.mm.1 |
NR_027372.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200010833.mm.1 |
NR_027372.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200010833.mm.1 |
NR_027373.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200010833.mm.1 |
NR_027373.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200010833.mm.1 |
TC0200001380.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200010834.mm.1 |
NM_001145902.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200010834.mm.1 |
NM_001145902.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200010834.mm.1 |
NM_146124.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200010834.mm.1 |
NM_146124.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200010834.mm.1 |
NR_027371.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200010834.mm.1 |
NR_027371.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200010834.mm.1 |
NR_027372.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200010834.mm.1 |
NR_027372.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200010834.mm.1 |
NR_027373.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200010834.mm.1 |
NR_027373.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200010834.mm.1 |
TC0200001380.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200010832.mm.1 |
NM_001145902.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200010832.mm.1 |
NM_001145902.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200010832.mm.1 |
NM_146124.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200010832.mm.1 |
NM_146124.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200010832.mm.1 |
NR_027371.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200010832.mm.1 |
NR_027371.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200010832.mm.1 |
NR_027372.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200010832.mm.1 |
NR_027372.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200010832.mm.1 |
NR_027373.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200010832.mm.1 |
NR_027373.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200010832.mm.1 |
TC0200001380.mm.1 |
MTA TC |