Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldh1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027406.1 | NP_081682.1 | 9 - 10 | 1221 | 72 | |
GenBank mRNA | BC024055.1 | - | 9 - 10 | 1178 | 5098789 | 72 |
BC025939.1 | - | 9 - 10 | 1203 | 5099360 | 72 | |
BC028817.1 | - | 9 - 10 | 1168 | 5101815 | 72 | |
BC030722.1 | - | 9 - 10 | 1205 | 5098671 | 72 | |
BC030723.1 | - | 9 - 10 | 1137 | 5100007 | 72 | |
BC030727.1 | - | 9 - 10 | 1207 | 5099026 | 72 | |
BC030730.1 | - | 9 - 10 | 1221 | 5101835 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |