Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007763.3 | NP_031789.1 | 1 - 2 | 142 | 98 | |
GenBank mRNA | AA266159.1 | - | 1 - 2 | 142 | 98 | |
AI323004.1 | - | 1 - 2 | 90 | 98 | ||
AK003075.1 | - | 1 - 2 | 104 | 98 | ||
AK008269.1 | - | 1 - 2 | 104 | 98 | ||
AK012068.2 | - | 1 - 2 | 99 | 98 | ||
AK088267.1 | - | 1 - 2 | 104 | 98 | ||
AK168305.1 | - | 1 - 2 | 103 | 98 | ||
BC031922.1 | - | 1 - 2 | 87 | 4206322 | 98 | |
BC064074.1 | - | 1 - 2 | 91 | 948562 | 98 | |
BX511593.1 | - | 1 - 2 | 88 | 98 | ||
DT908028.1 | - | 1 - 2 | 142 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |