Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01951665_u1
          See other SNRNP35 GE Assays ›
          Gene Symbol:
          Snrnp35
          Gene Name:
          small nuclear ribonucleoprotein 35 (U11/U12)
          Gene Aliases:
          6330548G22Rik
          Chromosome Location:
          Chr.5: 124483080 - 124491323 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01951665_u1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Snrnp35
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_029532.2 NP_083808.1 2 240 152
          GenBank mRNA AK018232.1 - 2 215 152
          AK039064.1 - 2 262 152
          BC043031.1 - 2 240 5693273 152
          BC058361.1 - 2 240 5693273 152

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Snrnp35

          Panther Classification:

          Molecular Function -

          RNA splicing factor

          Gene Ontology Categories:

          Function(s) Process(es)

          mRNA processing
          biological_process
          RNA splicing
          nucleotide binding
          molecular_function
          nucleic acid binding
          RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24061507 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24061510 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24061508 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1976690 BC043031.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1976690 BC058361.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1976690 NM_029532.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1976690 AK018232.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1976689 AK018232.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 105303 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 266966 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 130722 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 793171 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 166055 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P101488 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1976690 BC043031.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1976690 BC058361.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1976690 NM_029532.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1976690 AK018232.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1976689 AK018232.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24061513 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24061508 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24061517 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24061515 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24061510 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24061507 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24061518 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24061511 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P101488 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01211009 - -
          Whole Mouse Genome G4122A A_51_P441618 BC043031.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P441618 BC058361.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P441618 NM_029532.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P441618 AK018232.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P118994 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P101488 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04621659 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24061510 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24061508 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24061507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24061521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24061499 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24061493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24061526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24061480 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24061496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24061466 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24061489 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24061506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24061539 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24061503 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24061482 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24061486 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24061519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24061501 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24061511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24061557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24061476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24061558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24061536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24061464 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24061552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24061554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24061523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24061545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24061508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24061505 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24061509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24061472 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24061542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24061477 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24061485 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24061467 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24061524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24061481 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24061517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24061543 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24061553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24061470 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24061528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24061498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24061513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24061515 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24061517 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24061523 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24061513 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24061503 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24061539 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24061498 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24061506 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24061524 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24061542 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24061547 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04575685 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24061499 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04479009 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24061534 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24061511 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04266744 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04110497 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04353039 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24061509 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24061521 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04816669 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24061519 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24061501 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24061545 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04110494 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24061536 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24061528 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24061505 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10525742 - -
          Affymetrix Mouse Gene 1.0 ST Array 10525743 - -
          Affymetrix HT MG-430 PM Array Plate 1430041_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10525741 - -
          Mouse Genome 430 2.0 Array 1430041_at - -
          Mouse Genome 430 2.0 Array 1430041_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 76167_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 76167_at - -
          Affymetrix Mouse Clariom S Array PSR0500012967.mm.1 TC0500001445.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500012967.mm.1 NM_029532.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500012967.mm.1 NM_029532.2 Refseq
          Mouse Expression Set 430 1430041_PM_at - -
          Mouse Expression Set 430 1430041_at - -
          Affymetrix HT Mouse Genome 430B Array 1430041_at - -
          Affymetrix HT Mouse Genome 430B Array 1430041_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 664476 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3800315 - -
          Illumina mouse-6 v1.0 expression beadchip 3800315 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003800315 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2683844 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2683844 - -
          mouse-6 v1.1 (Illumina) scl27219.2.1_88-S - -
          Illumina Sentrix mouse-6.v1 bead array 3800315 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl27219.2.1_88-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3800315 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2935389 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2935386 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2935386 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2935389 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0