Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500007054.mm.1 |
TC0500000802.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500007054.mm.1 |
NM_007786.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500007054.mm.1 |
NM_007786.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500007048.mm.1 |
TC0500000802.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500007048.mm.1 |
NM_007786.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500007048.mm.1 |
NM_007786.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500007055.mm.1 |
TC0500000802.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500007055.mm.1 |
NM_007786.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500007055.mm.1 |
NM_007786.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500007059.mm.1 |
TC0500000802.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500007049.mm.1 |
TC0500000802.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500007049.mm.1 |
NM_007786.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500007049.mm.1 |
NM_007786.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500007052.mm.1 |
TC0500000802.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500007052.mm.1 |
NM_007786.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500007052.mm.1 |
NM_007786.4 |
Refseq |