Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001010941.2 | NP_001010941.1 | 2 | 1447 | 151 | |
NM_008151.3 | NP_032177.1 | 1 | 1233 | 151 | ||
GenBank mRNA | AK140298.1 | - | 2 | 1448 | 151 | |
BC055746.1 | - | 1 | 1233 | 6415225 | 151 | |
D21061.1 | - | 2 | 1035 | 151 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |