Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Iqsec2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005475.2 | NP_001005475.1 | 8 - 9 | 2188 | 146 | |
NM_001114664.2 | NP_001108136.1 | 8 - 9 | 2952 | 146 | ||
GenBank mRNA | AB275281.1 | - | 8 - 9 | 2952 | 146 | |
AK220345.1 | - | 2 - 3 | 223 | 146 | ||
AY451401.1 | - | 8 - 9 | 2945 | 146 | ||
CB236789.1 | - | 138 | 146 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |