Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Gene 1.0 ST Array |
10447648 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10447903 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10447647 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10447902 |
- |
- |
Affymetrix Mouse Clariom S Array |
PSR1700013098.mm.1 |
NM_001013832.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700013098.mm.1 |
NM_001013832.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700013098.mm.1 |
TC1700001463.mm.1 |
MTA TC |
Mouse Expression Set 430 |
1427421_PM_at |
- |
- |
Mouse Expression Set 430 |
1420527_s_at |
- |
- |
Mouse Expression Set 430 |
1427421_at |
- |
- |
Mouse Expression Set 430 |
1422225_PM_s_at |
- |
- |
Mouse Expression Set 430 |
1420527_PM_s_at |
- |
- |
Mouse Expression Set 430 |
1422225_s_at |
- |
- |
Murine Genome U74v2 Set |
100358_PM_s_at |
- |
- |
Murine Genome U74v2 Set |
100358_s_at |
- |
- |
Murine Genome U74v2 Set |
100359_at |
- |
- |
Murine Genome U74v2 Set |
103058_f_at |
- |
- |
Murine Genome U74v2 Set |
103058_PM_f_at |
- |
- |
Murine Genome U74v2 Set |
100359_PM_at |
- |
- |
Murine Genome U74v2 Set |
137662_at |
- |
- |
Murine Genome U74v2 Set |
137662_PM_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1427421_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1422225_PM_s_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1420527_s_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1420527_PM_s_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1427421_PM_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1422225_s_at |
- |
- |