Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc5a10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033227.2 | NP_001028399.1 | 3 - 4 | 427 | 56 | |
GenBank mRNA | AK144033.1 | - | 3 - 4 | 428 | 56 | |
AK165337.1 | - | 2 - 3 | 223 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |