Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028384.1 | NP_082660.1 | 4 - 5 | 569 | 93 | |
GenBank mRNA | AK158659.1 | - | 2 - 3 | 349 | 93 | |
BC147354.1 | - | 2 - 3 | 314 | 9056407 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |