Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl22l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026517.2 | NP_080793.1 | 2 - 3 | 127 | 105 | |
GenBank mRNA | AK008915.1 | - | 2 - 3 | 129 | 105 | |
AK013966.1 | - | 2 - 3 | 127 | 105 | ||
BC026533.1 | - | 2 - 3 | 107 | 5030430 | 105 | |
BC120716.1 | - | 2 - 3 | 202 | 40129639 | 105 | |
BC120718.1 | - | 2 - 3 | 202 | 40129641 | 105 |
Set Membership: |
Detect Genomic DNA Probe spans exons |