Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0800005242.mm.1 |
NM_001163282.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800005242.mm.1 |
NM_138647.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800005242.mm.1 |
NM_138647.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800005242.mm.1 |
TC0800000745.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800005241.mm.1 |
NM_001163282.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800005241.mm.1 |
NM_138647.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800005241.mm.1 |
NM_138647.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800005241.mm.1 |
TC0800000745.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800005247.mm.1 |
NM_001163282.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800005247.mm.1 |
NM_138647.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800005247.mm.1 |
NM_138647.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800005247.mm.1 |
TC0800000745.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800005249.mm.1 |
NM_001163282.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800005249.mm.1 |
NM_138647.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800005249.mm.1 |
NM_138647.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800005249.mm.1 |
TC0800000745.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800005253.mm.1 |
NM_001163282.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800005253.mm.1 |
NM_008107.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800005253.mm.1 |
NM_008107.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800005253.mm.1 |
NM_138647.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800005253.mm.1 |
NM_138647.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800005253.mm.1 |
TC0800000745.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800005255.mm.1 |
NM_001163282.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800005255.mm.1 |
NM_008107.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800005255.mm.1 |
NM_008107.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800005255.mm.1 |
NM_138647.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800005255.mm.1 |
NM_138647.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800005255.mm.1 |
TC0800000745.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800005256.mm.1 |
NM_001163282.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800005256.mm.1 |
NM_008107.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800005256.mm.1 |
NM_008107.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800005256.mm.1 |
NM_138647.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800005256.mm.1 |
NM_138647.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800005256.mm.1 |
TC0800000745.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800005246.mm.1 |
NM_001163282.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800005246.mm.1 |
NM_138647.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800005246.mm.1 |
NM_138647.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800005246.mm.1 |
TC0800000745.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800005254.mm.1 |
NM_001163282.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800005254.mm.1 |
NM_008107.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800005254.mm.1 |
NM_008107.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800005254.mm.1 |
NM_138647.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800005254.mm.1 |
NM_138647.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800005254.mm.1 |
TC0800000745.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800005243.mm.1 |
NM_001163282.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800005243.mm.1 |
NM_138647.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800005243.mm.1 |
NM_138647.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800005243.mm.1 |
TC0800000745.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800005252.mm.1 |
NM_001163282.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800005252.mm.1 |
NM_138647.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800005252.mm.1 |
NM_138647.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800005252.mm.1 |
TC0800000745.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800005248.mm.1 |
NM_001163282.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800005248.mm.1 |
NM_138647.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800005248.mm.1 |
NM_138647.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800005248.mm.1 |
TC0800000745.mm.1 |
MTA TC |