Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Gene 1.0 ST Array |
10604639 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10604634 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10604638 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10604640 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10604635 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10604636 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10604637 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10604633 |
- |
- |
Mouse Genome 430 2.0 Array |
1423816_PM_at |
- |
- |
Mouse Genome 430 2.0 Array |
1423816_at |
- |
- |
Affymetrix Mouse Clariom S Array |
PSR0X00014228.mm.1 |
NM_024170.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00014228.mm.1 |
NM_024170.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00014228.mm.1 |
TC0X00002241.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00014227.mm.1 |
NM_024170.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00014227.mm.1 |
NM_024170.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00014227.mm.1 |
TC0X00002241.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00014230.mm.1 |
NM_001018063.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00014230.mm.1 |
NM_001018063.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00014230.mm.1 |
TC0X00002242.mm.1 |
MTA TC |
Mouse Expression Set 430 |
1423816_PM_at |
- |
- |
Mouse Expression Set 430 |
1423816_at |
- |
- |
Murine Genome U74v2 Set |
166567_f_at |
- |
- |
Murine Genome U74v2 Set |
166567_PM_f_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1423816_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1423816_PM_at |
- |
- |