Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lhfpl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081231.2 | NP_001074700.2 | 3 - 4 | 719 | 143 | |
GenBank mRNA | AK033906.1 | - | 3 - 4 | 693 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mus musculus Probe spans exons |