Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100016281.mm.1 |
NM_001082535.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100016281.mm.1 |
NM_001082535.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100016281.mm.1 |
NM_199019.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100016281.mm.1 |
NM_199019.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100016281.mm.1 |
TC1100001724.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100016285.mm.1 |
NM_001082535.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100016285.mm.1 |
NM_001082535.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100016285.mm.1 |
TC1100001724.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100016282.mm.1 |
NM_001082535.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100016282.mm.1 |
NM_001082535.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100016282.mm.1 |
NM_199019.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100016282.mm.1 |
NM_199019.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100016282.mm.1 |
TC1100001724.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100016284.mm.1 |
NM_001082535.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100016284.mm.1 |
NM_001082535.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100016284.mm.1 |
TC1100001724.mm.1 |
MTA TC |