Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankfn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080933.1 | NP_001074402.1 | 1 - 2 | 143 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |