Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ero1lb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026184.2 | NP_080460.2 | 10 - 11 | 904 | 140 | |
GenBank mRNA | AF489856.1 | - | 10 - 11 | 710 | 140 | |
AK004981.1 | - | 881 | 140 | |||
AK006886.1 | - | 10 - 11 | 767 | 140 | ||
AK041951.1 | - | 10 - 11 | 880 | 140 | ||
AK160664.1 | - | 10 - 11 | 920 | 140 | ||
BC058721.1 | - | 10 - 11 | 904 | 5705507 | 140 | |
BC111900.1 | - | 10 - 11 | 874 | 40062137 | 140 | |
BC112426.1 | - | 10 - 11 | 874 | 40062136 | 140 | |
BC145888.1 | - | 10 - 11 | 855 | 40131191 | 140 | |
BC145890.1 | - | 10 - 11 | 855 | 40131193 | 140 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |