Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700012867.mm.1 |
TC0700001520.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700012867.mm.1 |
NM_001243916.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700012870.mm.1 |
TC0700001520.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700012870.mm.1 |
NM_001243916.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700012847.mm.1 |
TC0700001520.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700012847.mm.1 |
NM_030684.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700012847.mm.1 |
NM_030684.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700012861.mm.1 |
TC0700001520.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700012861.mm.1 |
NM_001243916.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700012848.mm.1 |
TC0700001520.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700012848.mm.1 |
NM_030684.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700012848.mm.1 |
NM_030684.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700012862.mm.1 |
TC0700001520.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700012862.mm.1 |
NM_001243916.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700012841.mm.1 |
TC0700001520.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700012841.mm.1 |
NM_030684.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700012841.mm.1 |
NM_030684.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700012842.mm.1 |
TC0700001520.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700012842.mm.1 |
NM_030684.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700012842.mm.1 |
NM_030684.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700012868.mm.1 |
TC0700001520.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700012868.mm.1 |
NM_001243916.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700012863.mm.1 |
TC0700001520.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700012863.mm.1 |
NM_001243916.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700012849.mm.1 |
TC0700001520.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700012849.mm.1 |
NM_030684.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700012849.mm.1 |
NM_030684.3 |
Refseq |