Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim34a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030684.3 | NP_109609.2 | 4 - 5 | 1079 | 73 | |
GenBank mRNA | AF220139.1 | - | 4 - 5 | 976 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |