Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rad23b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009011.4 | NP_033037.2 | 7 - 8 | 1137 | 74 | |
GenBank mRNA | AK142280.1 | - | 7 - 8 | 1129 | 74 | |
AK150089.1 | - | 7 - 8 | 1129 | 74 | ||
AK157252.1 | - | 7 - 8 | 1099 | 74 | ||
AK160880.1 | - | 7 - 8 | 1129 | 74 | ||
AK160890.1 | - | 7 - 8 | 1115 | 74 | ||
AK160973.1 | - | 7 - 8 | 1115 | 74 | ||
AK167582.1 | - | 7 - 8 | 1125 | 74 | ||
AK200838.1 | - | 3 - 4 | 304 | 74 | ||
AK218725.1 | - | 3 - 4 | 301 | 74 | ||
BC027747.1 | - | 7 - 8 | 1126 | 4459657 | 74 | |
BC068193.1 | - | 7 - 8 | 1096 | 6813997 | 74 | |
X92411.1 | - | 7 - 8 | 817 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |