Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm14342 | |||||||
---|---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | DQ860414.1 | - | LIT3342 HAR1R |
1 | 253 | 130 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
basic helix-loop-helix family, member e23 |
Bhlhe23 |
A930001L02Rik, BETA4, Bhlhb4 |
-60.9kb | Sense | Chr. 2 - 180774381 - 180776900 on NCBI Build GRCm38 |
Downstream | microRNA 124a-3 | Mir124a-3 |
Mirn124a-3, mir-124-3, mir-124a-3 |
55.4kb | Anti-sense | Chr. 2 - 180894040 - 180894107 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |