Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100011989.mm.1 |
NR_002840.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011989.mm.1 |
NR_002840.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011989.mm.1 |
TC0100001464.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100011996.mm.1 |
NR_002840.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011996.mm.1 |
NR_002840.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011996.mm.1 |
TC0100001464.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100012006.mm.1 |
NR_002840.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012006.mm.1 |
NR_002840.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012006.mm.1 |
TC0100001464.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100012011.mm.1 |
NR_002840.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012011.mm.1 |
NR_002840.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012011.mm.1 |
TC0100001464.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100012012.mm.1 |
NR_002840.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012012.mm.1 |
NR_002840.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012012.mm.1 |
TC0100001464.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100011997.mm.1 |
NR_002840.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011997.mm.1 |
NR_002840.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011997.mm.1 |
TC0100001464.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100012015.mm.1 |
NR_002840.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012015.mm.1 |
NR_002840.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012015.mm.1 |
TC0100001464.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100011994.mm.1 |
NR_002840.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011994.mm.1 |
NR_002840.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011994.mm.1 |
TC0100001464.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100012004.mm.1 |
NR_002840.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012004.mm.1 |
NR_002840.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012004.mm.1 |
TC0100001464.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100012010.mm.1 |
NR_002840.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012010.mm.1 |
NR_002840.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012010.mm.1 |
TC0100001464.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100011988.mm.1 |
NR_002840.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011988.mm.1 |
NR_002840.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011988.mm.1 |
TC0100001464.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100011990.mm.1 |
NR_002840.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011990.mm.1 |
NR_002840.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011990.mm.1 |
TC0100001464.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100011991.mm.1 |
NR_002840.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011991.mm.1 |
NR_002840.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011991.mm.1 |
TC0100001464.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100012003.mm.1 |
NR_002840.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012003.mm.1 |
NR_002840.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012003.mm.1 |
TC0100001464.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100012005.mm.1 |
NR_002840.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012005.mm.1 |
NR_002840.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012005.mm.1 |
TC0100001464.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100012007.mm.1 |
NR_002840.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012007.mm.1 |
NR_002840.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012007.mm.1 |
TC0100001464.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100011986.mm.1 |
NR_002840.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011986.mm.1 |
NR_002840.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011986.mm.1 |
TC0100001464.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100012019.mm.1 |
NR_002840.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012019.mm.1 |
NR_002840.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012019.mm.1 |
TC0100001464.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100011987.mm.1 |
NR_002840.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011987.mm.1 |
NR_002840.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011987.mm.1 |
TC0100001464.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100012000.mm.1 |
NR_002840.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012000.mm.1 |
NR_002840.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012000.mm.1 |
TC0100001464.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100012001.mm.1 |
NR_002840.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012001.mm.1 |
NR_002840.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012001.mm.1 |
TC0100001464.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100011992.mm.1 |
NR_002840.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011992.mm.1 |
NR_002840.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011992.mm.1 |
TC0100001464.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100011995.mm.1 |
NR_002840.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011995.mm.1 |
NR_002840.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011995.mm.1 |
TC0100001464.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100011998.mm.1 |
NR_002840.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011998.mm.1 |
NR_002840.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011998.mm.1 |
TC0100001464.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100012020.mm.1 |
NR_002840.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012020.mm.1 |
NR_002840.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012020.mm.1 |
TC0100001464.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100012029.mm.1 |
NR_002840.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012029.mm.1 |
NR_002840.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012029.mm.1 |
TC0100001464.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100011985.mm.1 |
NR_002840.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011985.mm.1 |
NR_002840.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011985.mm.1 |
TC0100001464.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100011999.mm.1 |
NR_002840.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011999.mm.1 |
NR_002840.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011999.mm.1 |
TC0100001464.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100012008.mm.1 |
NR_002840.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012008.mm.1 |
NR_002840.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012008.mm.1 |
TC0100001464.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100012017.mm.1 |
NR_002840.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012017.mm.1 |
NR_002840.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012017.mm.1 |
TC0100001464.mm.1 |
MTA TC |