Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1700018H16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK006090.1 | - | 1 | 342 | 95 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | WAP four-disulfide core domain 18 | Wfdc18 |
Expi, Kal1, WDNM1 |
-33.1kb | Sense | Chr. 11 - 83709004 - 83711360 on NCBI Build GRCm38 |
Downstream | WAP four-disulfide core domain 21 | Wfdc21 |
1100001G20Rik, Wdnm1-like |
2.1kb | Sense | Chr. 11 - 83746940 - 83752646 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |