Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4930477E14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK015579.1 | - | 1 - 2 | 138 | 71 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | U1b2 small nuclear RNA | Rnu1b2 | pU1.2 | -27.0kb | Anti-sense | Chr. 3 - 96374021 - 96374185 on NCBI Build GRCm38 |
Downstream | telomerase RNA component | Terc |
mTER, mTR |
12.4kb | Sense | Chr. 3 - 96414437 - 96414833 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus 1700 Probe spans exons |