Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | neuropeptide Y receptor Y2 | Npy2r | NPY2-R | Sense | Chr. 3 - 82538383 - 82548989 on NCBI Build GRCm38 | |
Downstream |
predicted gene, 29999 |
Gm29999 | 116.0kb | Sense | Chr. 3 - 82660521 - 82664900 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |