Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | orthodenticle homolog 2 opposite strand 1 | Otx2os1 | -86.2kb | Sense | Chr. 14 - 48669288 - 48793348 on NCBI Build GRCm38 | |
Downstream |
predicted gene, 36024 |
Gm36024 | 102.4kb | Anti-sense | Chr. 14 - 48982480 - 48984380 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |