Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | heterogeneous nuclear ribonucleoprotein A3 | Hnrnpa3 |
2410013L13Rik, 2610209F03Rik, 2610510D13Rik, Hnrpa3 |
-14.0kb | Anti-sense | Chr. 2 - 75659230 - 75669407 on NCBI Build GRCm38 |
Downstream | RIKEN cDNA E030042O20 gene | E030042O20Rik | 20.8kb | Anti-sense | Chr. 2 - 75704770 - 75711120 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |