Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | nerve growth factor | Ngf | Ngfb | Sense | Chr. 3 - 102469919 - 102521013 on NCBI Build GRCm38 | |
Downstream |
tetraspanin 2, opposite strand |
Tspan2os | 2410057H14Rik | 226.0kb | Anti-sense | Chr. 3 - 102720231 - 102735417 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |