Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Triml2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001160412.1 | NP_001153884.1 | 4 - 5 | 659 | 107 | |
GenBank mRNA | AV448075.1 | - | 2 - 3 | 162 | 107 | |
EU599038.1 | - | 6 - 7 | 749 | 107 | ||
EU599039.1 | - | 5 - 6 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |