Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100021565.mm.1 |
NM_001163359.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100021565.mm.1 |
NM_001163359.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100021565.mm.1 |
NM_001163360.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100021565.mm.1 |
NM_001163360.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100021565.mm.1 |
NM_021891.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100021565.mm.1 |
NM_021891.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100021565.mm.1 |
TC1100002224.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100021562.mm.1 |
NM_001163359.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100021562.mm.1 |
NM_001163359.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100021562.mm.1 |
NM_001163360.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100021562.mm.1 |
NM_001163360.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100021562.mm.1 |
NM_021891.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100021562.mm.1 |
NM_021891.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100021562.mm.1 |
TC1100002224.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100021564.mm.1 |
NM_001163359.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100021564.mm.1 |
NM_001163359.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100021564.mm.1 |
NM_001163360.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100021564.mm.1 |
NM_001163360.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100021564.mm.1 |
NM_021891.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100021564.mm.1 |
NM_021891.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100021564.mm.1 |
TC1100002224.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100021559.mm.1 |
NM_001163359.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100021559.mm.1 |
NM_001163359.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100021559.mm.1 |
NM_001163360.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100021559.mm.1 |
NM_001163360.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100021559.mm.1 |
NM_021891.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100021559.mm.1 |
NM_021891.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100021559.mm.1 |
TC1100002224.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100021563.mm.1 |
NM_001163359.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100021563.mm.1 |
NM_001163359.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100021563.mm.1 |
NM_001163360.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100021563.mm.1 |
NM_001163360.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100021563.mm.1 |
NM_021891.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100021563.mm.1 |
NM_021891.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100021563.mm.1 |
TC1100002224.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100021560.mm.1 |
NM_001163359.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100021560.mm.1 |
NM_001163359.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100021560.mm.1 |
NM_001163360.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100021560.mm.1 |
NM_001163360.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100021560.mm.1 |
NM_021891.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100021560.mm.1 |
NM_021891.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100021560.mm.1 |
TC1100002224.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100021561.mm.1 |
NM_001163359.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100021561.mm.1 |
NM_001163359.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100021561.mm.1 |
NM_001163360.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100021561.mm.1 |
NM_001163360.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100021561.mm.1 |
NM_021891.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100021561.mm.1 |
NM_021891.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100021561.mm.1 |
TC1100002224.mm.1 |
MTA TC |