Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zswim7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027198.1 | NP_081474.1 | 2 - 3 | 166 | 60 | |
GenBank mRNA | AK010472.1 | - | 2 - 3 | 146 | 60 | |
BC109343.2 | - | 2 - 3 | 124 | 40053371 | 60 | |
BQ554850.1 | - | 2 - 3 | 169 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |