Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam160b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_194345.1 | NP_919326.1 | 15 - 16 | 2042 | 70 | |
GenBank mRNA | AK090035.1 | - | 10 - 11 | 1294 | 70 | |
BC037228.1 | - | 3 - 4 | 319 | 70 | ||
BC050861.1 | - | 14 - 15 | 2165 | 70 | ||
BC058108.1 | - | 3 - 4 | 347 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |