Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0800010445.mm.1 |
NM_001039700.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800010445.mm.1 |
NM_001039700.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800010445.mm.1 |
NM_001286454.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800010445.mm.1 |
NM_181544.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800010445.mm.1 |
NM_181544.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800010445.mm.1 |
TC0800001298.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800010444.mm.1 |
NM_001039700.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800010444.mm.1 |
NM_001039700.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800010444.mm.1 |
NM_001286454.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800010444.mm.1 |
NM_181544.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800010444.mm.1 |
NM_181544.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800010444.mm.1 |
TC0800001298.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800010446.mm.1 |
NM_001039700.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800010446.mm.1 |
NM_001039700.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800010446.mm.1 |
NM_001286454.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800010446.mm.1 |
NM_181544.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800010446.mm.1 |
NM_181544.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800010446.mm.1 |
TC0800001298.mm.1 |
MTA TC |